Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145724.1 | NP_663776.1 | 2 | 592 | ||
GenBank mRNA | AF052042.1 | - | 2 | 592 | ||
BC100140.1 | - | 2 | 586 | 7442414 |