Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBA52 SIRNA Assays ›
          Gene Symbol:
          Uba52
          Gene Name:
          ubiquitin A-52 residue ribosomal protein fusion product 1
          Gene Aliases:
          Rps27a, Ubb
          Chromosome Location:
          Chr.16: 20668941 - 20671127 on Build Rnor_6.0
          UniGene ID:
          Rn.110618
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 59369

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Uba52
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031687.3 NP_113875.1 2, 3 144
          XM_006252934.2 XP_006252996.1 2, 3 231
          XM_006252935.1 XP_006252997.1 2, 3 349
          XM_008771145.1 XP_008769367.1 2, 3 227
          XM_008771146.1 XP_008769368.1 2, 3 171
          GenBank mRNA BC061544.1 - 2, 3 140 6922034
          BP492585.1 - 1, 2 30
          FQ209944.1 - 2, 3 136
          FQ210541.1 - 2, 3 133
          FQ210904.1 - 2, 3 134
          FQ212001.1 - 2, 3 138
          FQ212205.1 - 2, 3 136
          FQ212291.1 - 2, 3 134
          FQ212810.1 - 2, 3 137
          FQ217440.1 - 2, 3 129
          FQ218121.1 - 2, 3 130
          FQ219376.1 - 2, 3 135
          FQ219803.1 - 2, 3 136
          FQ221045.1 - 2, 3 133
          FQ221118.1 - 2, 3 134
          FQ221238.1 - 2, 3 354
          FQ221621.1 - 2, 3 130
          FQ221631.1 - 2, 3 136
          FQ221755.1 - 2, 3 134
          FQ221772.1 - 2, 3 134
          FQ221777.1 - 2, 3 134
          FQ222045.1 - 2, 3 134
          FQ222098.1 - 2, 3 130
          FQ222371.1 - 2, 3 133
          FQ222440.1 - 2, 3 145
          FQ222483.1 - 2, 3 133
          FQ222507.1 - 2, 3 128
          FQ222550.1 - 2, 3 130
          FQ222700.1 - 2, 3 133
          FQ223258.1 - 2, 3 136
          FQ223423.1 - 2, 3 130
          FQ223456.1 - 2, 3 131
          FQ224647.1 - 1, 2 124
          FQ224702.1 - 2, 3 134
          FQ228362.1 - 2, 3 130
          FQ228566.1 - 2, 3 134
          X82636.1 - 1, 2 106

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_021009, AB009010, BC093445, AF052164, M26880, and others.
          This siRNA also targets the following transcripts in Mouse: NM_019883, EF437347, AK018726, AF118402, CK620747, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0