Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053800.3 | NP_446252.1 | 1, 2 | 125 | ||
GenBank mRNA | AF311055.1 | - | 1, 2 | 125 | ||
BC058454.1 | - | 210 | 6886988 | |||
FQ209943.1 | - | 207 | ||||
FQ210839.1 | - | 210 | ||||
FQ216849.1 | - | 236 | ||||
FQ217822.1 | - | 205 | ||||
FQ221646.1 | - | 178 | ||||
X14878.1 | - | 185 |