Search
Search

DAPK1| Gene Symbol: DAPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288729.1 | NP_001275658.1 | 16, 17 | 1844 | ||
| NM_001288730.1 | NP_001275659.1 | 16, 17 | 1834 | |||
| NM_001288731.1 | NP_001275660.1 | 16, 17 | 1817 | |||
| NM_004938.3 | NP_004929.2 | 16, 17 | 2169 | |||
| XM_005251757.3 | XP_005251814.1 | 16, 17 | 1939 | |||
| XM_017014360.1 | XP_016869849.1 | 3, 4 | 1082 | |||
| GenBank mRNA | AB208871.1 | - | 16, 17 | 1772 | ||
| AK127855.1 | - | 2, 3 | 2260 | |||
| AK295119.1 | - | 16, 17 | 1901 | |||
| BC113660.1 | - | 15, 16 | 1707 | 8322712 | ||
| BC143730.1 | - | 14, 15 | 1657 | |||
| BC143733.1 | - | 15, 16 | 1707 | 9052247 | ||
| BC143759.1 | - | 15, 16 | 1707 | 9052273 | ||
| CR749834.1 | - | 16, 17 | 1817 | |||
| DA234399.1 | - | 3, 4 | 347 | |||
| X76104.1 | - | 16, 17 | 1973 | |||
Set Membership: |
|