Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007460.1 | NP_031486.1 | 19 | 2445 | ||
GenBank mRNA | AB004305.1 | - | 19 | 2445 | ||
AK155518.1 | - | 19 | 2495 | |||
AK181516.1 | - | 2 | 145 | |||
BC048786.1 | - | 19 | 2435 | 6489789 | ||
BC053066.1 | - | 19 | 2461 | 5704929 |