Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018235.1 | - | 1 | 411 | ||
AK031259.1 | - | 5 | 1174 | |||
AK042977.1 | - | 5 | 1214 | |||
AK053123.1 | - | 5 | 1632 | |||
AK077591.1 | - | 5 | 1187 | |||
AK167133.1 | - | 5 | 1184 | |||
AK167601.1 | - | 5 | 1188 | |||
AK167653.1 | - | 5 | 1187 | |||
AK169094.1 | - | 5 | 1184 | |||
AK169412.1 | - | 5 | 1184 | |||
BC010487.1 | - | 5 | 1467 | 3483748 |