Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1A SIRNA Assays ›
          Gene Symbol:
          Cacna1a
          Gene Name:
          calcium channel, voltage-dependent, P/Q type, alpha 1A subunit
          Gene Aliases:
          APCA, BI, Caca1a, Cacnl1a4, Cav2.1, Ccha1a, EA2, FHM, HPCA, MHP, MHP1, SCA6, alpha1A, la, nmf352, rkr, tg
          Chromosome Location:
          Chr.8: 84338633 - 84640249 on Build GRCm38
          UniGene ID:
          Mm.334658
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 60272

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cacna1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252059.1 NP_001238988.1 2 595
          NM_001252060.1 NP_001238989.1 3 736
          NM_001252061.1 NP_001238990.1 3 736
          NM_007578.3 NP_031604.3 3 736
          GenBank mRNA AB066608.1 - 3 509
          AB066609.1 - 3 509
          AK052625.1 - 4 539
          AK147277.1 - 2 583
          AK147288.1 - 3 736
          AK147498.1 - 2 583
          AK147697.1 - 2 583
          AY714490.1 - 3 509
          U76716.1 - 3 311

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012918, XM_008772355, XM_008772348, XM_008772359, XM_008772357, and others.

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          sulfur amino acid metabolic process
          glucose metabolic process
          transport
          ion transport
          calcium ion transport
          positive regulation of cytosolic calcium ion concentration
          gamma-aminobutyric acid signaling pathway
          chemical synaptic transmission
          neuron-neuron synaptic transmission
          neuromuscular synaptic transmission
          synapse assembly
          adult walking behavior
          cell death
          gamma-aminobutyric acid secretion
          regulation of acetylcholine secretion, neurotransmission
          cell growth
          calcium ion regulated exocytosis
          regulation of calcium ion-dependent exocytosis
          transmission of nerve impulse
          sensory perception of pain
          spinal cord motor neuron differentiation
          cerebellum maturation
          cerebellar molecular layer development
          cerebellar Purkinje cell layer development
          cerebellar Purkinje cell differentiation
          vestibular nucleus development
          central nervous system neuron differentiation
          cellular chloride ion homeostasis
          negative regulation of hormone biosynthetic process
          regulation of ion transmembrane transport
          synaptic transmission, glutamatergic
          neurotransmitter metabolic process
          regulation of membrane potential
          thyroid hormone metabolic process
          hormone metabolic process
          receptor clustering
          negative regulation of neuron apoptotic process
          response to pain
          behavioral response to pain
          calcium ion-regulated exocytosis of neurotransmitter
          dendrite morphogenesis
          regulation of axonogenesis
          neurological system process
          musculoskeletal movement, spinal reflex action
          neuromuscular process controlling balance
          neuromuscular process
          membrane depolarization
          synaptic transmission, GABAergic
          transmembrane transport
          rhythmic synaptic transmission
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          DNA binding
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated calcium channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          high voltage-gated calcium channel activity
          syntaxin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0