Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BDNF SIRNA Assays ›
          Gene Symbol:
          Bdnf
          Gene Name:
          brain derived neurotrophic factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 109674700 - 109727043 on Build GRCm38
          UniGene ID:
          Mm.1442
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 60424

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bdnf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001048139.1 NP_001041604.1 2 867
          NM_001048141.1 NP_001041606.1 2 707
          NM_001048142.1 NP_001041607.1 2 715
          NM_001285416.1 NP_001272345.1 2 784
          NM_001285417.1 NP_001272346.1 2 581
          NM_001285418.1 NP_001272347.1 2 572
          NM_001285419.1 NP_001272348.1 2 561
          NM_001285420.1 NP_001272349.1 2 449
          NM_001285421.1 NP_001272350.1 2 647
          NM_001285422.1 NP_001272351.1 1 829
          NM_007540.4 NP_031566.4 2 1008
          GenBank mRNA AK017559.1 - 2 649
          AK033127.1 - 2 661
          AY057908.1 - 2 1008
          AY057909.1 - 2 572
          AY057910.1 - 2 784
          AY057911.1 - 2 867
          AY057912.1 - 2 499
          AY057913.1 - 2 707
          AY057914.1 - 2 715
          AY231131.1 - 2 521
          AY231132.1 - 3 596
          BC034862.1 - 1 374 1397218
          EF125669.1 - 2 1008
          EF125670.1 - 2 573
          EF125671.1 - 2 785
          EF125672.1 - 2 869
          EF125673.1 - 2 707
          EF125674.1 - 2 715
          EF125681.1 - 2 561
          EF125682.1 - 2 449
          EF125683.1 - 2 581
          EF125684.1 - 2 647
          EF125685.1 - 1 826
          X55573.1 - 2 424

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001270633, NM_012513, NM_001270630, NM_001270631, NM_001270638, and others.

          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          behavioral fear response
          mitochondrial electron transport, NADH to ubiquinone
          transmembrane receptor protein tyrosine kinase signaling pathway
          gamma-aminobutyric acid signaling pathway
          cell-cell signaling
          nervous system development
          negative regulation of neuroblast proliferation
          axon guidance
          axon target recognition
          learning or memory
          learning
          feeding behavior
          neuron recognition
          glutamate secretion
          dendrite development
          regulation of metabolic process
          nerve development
          regulation of axon extension
          negative regulation of synaptic transmission, GABAergic
          positive regulation of peptidyl-serine phosphorylation
          mechanoreceptor differentiation
          response to drug
          fear response
          negative regulation of apoptotic process
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of neuron differentiation
          negative regulation of striated muscle tissue development
          regulation of retinal cell programmed cell death
          regulation of synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          positive regulation of long-term neuronal synaptic plasticity
          regulation of short-term neuronal synaptic plasticity
          axon extension
          neuron projection morphogenesis
          inner ear development
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of synapse assembly
          excitatory postsynaptic potential
          inhibitory postsynaptic potential
          negative regulation of cell death
          taste bud development
          cellular response to acid chemical
          dendrite extension
          negative regulation of neuron death
          neuron projection extension
          positive regulation of glucocorticoid receptor signaling pathway
          receptor binding
          neurotrophin TRKB receptor binding
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0