Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYP26A1 SIRNA Assays ›
          Gene Symbol:
          Cyp26a1
          Gene Name:
          cytochrome P450, family 26, subfamily a, polypeptide 1
          Gene Aliases:
          Cyp26, P450RA, P450RAI, RAH
          Chromosome Location:
          Chr.19: 37697808 - 37701536 on Build GRCm38
          UniGene ID:
          Mm.42230
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61021

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cyp26a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007811.2 NP_031837.2 7 1280
          GenBank mRNA AA239785.1 - 3 209
          AF115769.1 - 7 1204
          BC012673.1 - 7 1240 4210893
          Y12657.1 - 7 1234

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          central nervous system development
          anterior/posterior pattern specification
          neural crest cell development
          retinoic acid catabolic process
          retinoic acid metabolic process
          retinoic acid receptor signaling pathway
          oxidation-reduction process
          retinoic acid binding
          monooxygenase activity
          iron ion binding
          retinoic acid 4-hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          fluorene oxygenase activity
          mono-butyltin dioxygenase activity
          tri-n-butyltin dioxygenase activity
          di-n-butyltin dioxygenase activity
          methylsilanetriol hydroxylase activity
          methyl tertiary butyl ether 3-monooxygenase activity
          4-nitrocatechol 4-monooxygenase activity
          4-chlorophenoxyacetate monooxygenase activity
          tert-butanol 2-monooxygenase activity
          alpha-pinene monooxygenase activity
          dimethylsilanediol hydroxylase activity
          ammonia monooxygenase activity
          hydroxymethylsilanetriol oxidase activity
          2-hydroxyisobutyrate 3-monooxygenase activity
          alpha-pinene dehydrogenase activity
          heme binding
          bisphenol A hydroxylase B activity
          2,2-bis(4-hydroxyphenyl)-1-propanol hydroxylase activity
          9-fluorenone-3,4-dioxygenase activity
          anthracene 9,10-dioxygenase activity
          2-(methylthio)benzothiazole monooxygenase activity
          2-hydroxybenzothiazole monooxygenase activity
          benzothiazole monooxygenase activity
          2,6-dihydroxybenzothiazole monooxygenase activity
          pinacolone 5-monooxygenase activity
          thioacetamide S-oxygenase activity
          thioacetamide S-oxide S-oxygenase activity
          endosulfan monooxygenase I activity
          N-nitrodimethylamine hydroxylase activity
          4-(1-ethyl-1,4-dimethyl-pentyl)phenol monoxygenase activity
          endosulfan ether monooxygenase activity
          pyrene 4,5-monooxygenase activity
          pyrene 1,2-monooxygenase activity
          1-hydroxypyrene 6,7-monooxygenase activity
          1-hydroxypyrene 7,8-monooxygenase activity
          phenylboronic acid monooxygenase activity
          spheroidene monooxygenase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0