Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A10 SIRNA Assays ›
Gene Symbol:
SLC4A10
Gene Name:
solute carrier family 4 member 10
Gene Aliases:
NBCn2, NCBE
Chromosome Location:
Chr.2: 161624335 - 161985276 on Build GRCh38
UniGene ID:
Hs.333958
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6133
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178015.1
NP_001171486.1
2
271
NM_001178016.1
NP_001171487.1
3
407
NM_022058.3
NP_071341.2
2
271
XM_005246694.4
XP_005246751.1
3
326
XM_005246695.4
XP_005246752.1
3
326
XM_011511508.2
XP_011509810.1
2
114
XM_011511509.2
XP_011509811.1
2
114
XM_011511510.2
XP_011509812.1
2
114
XM_011511511.2
XP_011509813.1
2
114
XM_011511512.2
XP_011509814.1
2
114
XM_011511513.2
XP_011509815.1
5
603
XM_017004541.1
XP_016860030.1
2
114
XM_017004542.1
XP_016860031.1
2
114
XM_017004543.1
XP_016860032.1
2
114
XM_017004544.1
XP_016860033.1
2
114
XM_017004545.1
XP_016860034.1
4
512
XM_017004546.1
XP_016860035.1
5
603
XM_017004547.1
XP_016860036.1
3
326
XM_017004548.1
XP_016860037.1
3
300
XM_017004549.1
XP_016860038.1
4
478
XM_017004550.1
XP_016860039.1
3
326
XM_017004552.1
XP_016860041.1
2
114
XM_017004553.1
XP_016860042.1
2
114
GenBank mRNA
AB040457.1
-
2
179
AK293793.1
-
3
320
AK294767.1
-
2
208
AK308529.1
-
5
603
AK309839.1
-
3
326
AY376402.1
-
2
87
BC136269.1
-
2
157
9020256
BC143714.1
-
2
157
9052228
DC337667.1
-
2
272
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
chloride transport
nervous system development
response to light stimulus
post-embryonic development
bicarbonate transport
pyramidal neuron development
multicellular organism growth
locomotory exploration behavior
brain morphogenesis
regulation of intracellular pH
anion transmembrane transport
hydrogen ion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
symporter activity
anion:anion antiporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-nmj5c:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0