Search
Search

Fdx1| Gene Symbol: Fdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301728.1 | NP_001288657.1 | 3 | 515 | ||
| NM_007996.2 | NP_032022.1 | 3 | 515 | |||
| GenBank mRNA | AK004879.1 | - | 3 | 469 | ||
| AK148115.1 | - | 3 | 470 | |||
| BC099518.1 | - | 3 | 369 | |||
| BC132173.1 | - | 3 | 440 | 40130450 | ||
| BC132175.1 | - | 3 | 440 | 40130452 | ||
| D43689.1 | - | 3 | 446 | |||
| D43690.1 | - | 3 | 441 | |||
| L29123.1 | - | 3 | 468 | |||