Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164717.1 | NP_001158189.1 | 2 | 412 | ||
NM_008018.4 | NP_032044.2 | 2 | 412 | |||
GenBank mRNA | AJ007012.1 | - | 2 | 412 | ||
BC118022.1 | - | 2 | 248 | 40097448 | ||
CJ103307.1 | - | 2 | 424 |