Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF2 SIRNA Assays ›
          Gene Symbol:
          Fgf2
          Gene Name:
          fibroblast growth factor 2
          Gene Aliases:
          Fgf-2, Fgfb, bFGF
          Chromosome Location:
          Chr.3: 37348469 - 37405538 on Build GRCm38
          UniGene ID:
          Mm.473689
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61468

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fgf2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008006.2 NP_032032.1 3 634
          GenBank mRNA AF065903.1 - 3 437
          AF065904.1 - 3 437
          AF065905.1 - 3 437
          AY027548.1 - 3 352
          AY027549.1 - 3 457
          AY027550.1 - 1 229
          AY027551.1 - 3 434
          AY027557.1 - 3 507
          AY027558.1 - 3 498
          AY027559.1 - 3 361
          BY730691.1 - 3 612
          M30644.1 - 3 437

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019305, X07285, M22427.

          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          angiogenesis
          branching involved in ureteric bud morphogenesis
          organ induction
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          cell migration involved in sprouting angiogenesis
          phosphatidylinositol biosynthetic process
          multicellular organism development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          embryo development ending in birth or egg hatching
          glial cell differentiation
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of fibroblast migration
          positive regulation of phospholipase C activity
          growth factor dependent regulation of skeletal muscle satellite cell proliferation
          substantia nigra development
          positive regulation of cerebellar granule cell precursor proliferation
          cell differentiation
          hyaluronan catabolic process
          lung development
          inositol phosphate biosynthetic process
          wound healing
          positive regulation of cell fate specification
          positive regulation of MAP kinase activity
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of retinal cell programmed cell death
          positive regulation of smooth muscle cell proliferation
          response to axon injury
          stem cell development
          positive regulation of epithelial cell proliferation
          positive chemotaxis
          release of sequestered calcium ion into cytosol
          regulation of cell cycle
          positive regulation of cell division
          positive regulation of protein kinase B signaling
          cardiac muscle cell proliferation
          positive regulation of cardiac muscle cell proliferation
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          negative regulation of cell death
          chondroblast differentiation
          mammary gland epithelial cell differentiation
          angiogenesis involved in coronary vascular morphogenesis
          negative regulation of wound healing
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          positive regulation of sprouting angiogenesis
          regulation of endothelial cell chemotaxis to fibroblast growth factor
          positive regulation of endothelial cell chemotaxis to fibroblast growth factor
          receptor binding
          fibroblast growth factor receptor binding
          cytokine activity
          protein binding
          growth factor activity
          heparin binding
          ligand-dependent nuclear receptor transcription coactivator activity
          chemoattractant activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0