Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC4A10 SIRNA Assays ›
          Gene Symbol:
          SLC4A10
          Gene Name:
          solute carrier family 4 member 10
          Gene Aliases:
          NBCn2, NCBE
          Chromosome Location:
          Chr.2: 161624335 - 161985276 on Build GRCh38
          UniGene ID:
          Hs.333958
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 6229

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC4A10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178015.1 NP_001171486.1 2, 3 322
          NM_001178016.1 NP_001171487.1 3, 4 458
          NM_022058.3 NP_071341.2 2, 3 322
          XM_005246694.4 XP_005246751.1 3, 4 377
          XM_005246695.4 XP_005246752.1 3, 4 377
          XM_011511508.2 XP_011509810.1 2, 3 165
          XM_011511509.2 XP_011509811.1 2, 3 165
          XM_011511510.2 XP_011509812.1 2, 3 165
          XM_011511511.2 XP_011509813.1 2, 3 165
          XM_011511512.2 XP_011509814.1 2, 3 165
          XM_011511513.2 XP_011509815.1 5, 6 654
          XM_017004541.1 XP_016860030.1 2, 3 165
          XM_017004542.1 XP_016860031.1 2, 3 165
          XM_017004543.1 XP_016860032.1 2, 3 165
          XM_017004544.1 XP_016860033.1 2, 3 165
          XM_017004545.1 XP_016860034.1 4, 5 563
          XM_017004546.1 XP_016860035.1 5, 6 654
          XM_017004547.1 XP_016860036.1 3, 4 377
          XM_017004548.1 XP_016860037.1 3, 4 351
          XM_017004549.1 XP_016860038.1 4, 5 529
          XM_017004550.1 XP_016860039.1 3, 4 377
          XM_017004552.1 XP_016860041.1 2, 3 165
          XM_017004553.1 XP_016860042.1 2, 3 165
          GenBank mRNA AB040457.1 - 2, 3 230
          AK293793.1 - 3, 4 371
          AK294767.1 - 2, 3 259
          AK308529.1 - 5, 6 654
          AK309839.1 - 3, 4 377
          AY376402.1 - 2, 3 138
          BC136269.1 - 2, 3 208 9020256
          BC143714.1 - 2, 3 208 9052228

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          chloride transport
          nervous system development
          response to light stimulus
          post-embryonic development
          bicarbonate transport
          pyramidal neuron development
          multicellular organism growth
          locomotory exploration behavior
          brain morphogenesis
          regulation of intracellular pH
          anion transmembrane transport
          hydrogen ion transmembrane transport
          inorganic anion exchanger activity
          anion transmembrane transporter activity
          symporter activity
          anion:anion antiporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0