Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il12rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008353.2 | NP_032379.2 | 8 | 891 | ||
GenBank mRNA | AK137577.1 | - | 8 | 892 | ||
AK171937.1 | - | 8 | 868 | |||
BC057212.1 | - | 8 | 874 | 5350447 | ||
U23922.1 | - | 8 | 846 |