Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008550.2 | NP_032576.2 | 1, 2 | 220 | ||
GenBank mRNA | AB006458.1 | - | 1, 2 | 198 | ||
AK004900.1 | - | 1, 2 | 174 | |||
AK040388.1 | - | 1, 2 | 181 | |||
AK079969.1 | - | 1, 2 | 201 | |||
AK151873.1 | - | 1, 2 | 200 | |||
AK152349.1 | - | 1, 2 | 200 | |||
AK165376.1 | - | 1, 2 | 213 | |||
BC066211.1 | - | 1, 2 | 171 | 30101816 | ||
BY315451.1 | - | 1, 2 | 225 |