Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOG SIRNA Assays ›
Gene Symbol:
Nog
Gene Name:
noggin
Gene Aliases:
-
Chromosome Location:
Chr.11: 89300638 - 89302559 on Build GRCm38
UniGene ID:
Mm.135266
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
63422
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nog
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008711.2
NP_032737.1
1
975
GenBank mRNA
AK146488.1
-
1
978
BC132527.1
-
1
501
40130804
BC138148.1
-
1
501
8861167
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_005450, BC034027, AK314080.
This siRNA also targets the following transcripts in Rat: NM_012990, U31203.
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
osteoblast differentiation
urogenital system development
ureteric bud development
in utero embryonic development
endoderm formation
mesoderm formation
epithelial to mesenchymal transition
neural plate morphogenesis
neural tube closure
multicellular organism development
pattern specification process
axon guidance
central nervous system development
brain development
motor neuron axon guidance
dorsal/ventral pattern formation
negative regulation of gene expression
spinal cord development
cell differentiation in hindbrain
neural tube development
pituitary gland development
cell differentiation
negative regulation of cell migration
BMP signaling pathway
regulation of BMP signaling pathway
negative regulation of BMP signaling pathway
somatic stem cell population maintenance
wound healing
middle ear morphogenesis
embryonic digit morphogenesis
negative regulation of cell differentiation
regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
axial mesoderm development
notochord morphogenesis
anatomical structure formation involved in morphogenesis
embryonic skeletal system development
negative regulation of astrocyte differentiation
mesenchymal cell differentiation
positive regulation of epithelial cell proliferation
cartilage development
negative regulation of cardiac muscle cell proliferation
limb development
embryonic skeletal joint morphogenesis
negative regulation of cytokine activity
face morphogenesis
negative regulation of pathway-restricted SMAD protein phosphorylation
lung morphogenesis
prostatic bud formation
ureteric bud formation
fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
negative regulation of cartilage development
somite development
cellular response to BMP stimulus
negative regulation of canonical Wnt signaling pathway
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of glomerulus development
regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
negative regulation of apoptotic signaling pathway
cytokine binding
protein complex binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0