Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008943.2 | NP_032969.1 | 4 | 682 | ||
GenBank mRNA | AK134232.1 | - | 4 | 365 | ||
AK134615.1 | - | 4 | 404 | |||
AK165984.1 | - | 4 | 403 | |||
BC014744.1 | - | 4 | 433 | 4511140 | ||
BC030409.1 | - | 4 | 357 | 5133302 | ||
BC071233.1 | - | 4 | 421 | 3673609 | ||
L42177.1 | - | 4 | 378 |