Search Thermo Fisher Scientific
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159729.1 | NP_001153201.1 | 3 | 425 | ||
NM_008999.4 | NP_033025.2 | 3 | 537 | |||
GenBank mRNA | AB232618.1 | - | 1 | 102 | ||
AK016493.1 | - | 3 | 437 | |||
AK045454.1 | - | 3 | 296 | |||
AK047068.1 | - | 3 | 538 | |||
AK054079.1 | - | 3 | 523 | |||
AK082017.1 | - | 3 | 500 | |||
AK163909.1 | - | 3 | 546 | |||
BC025578.1 | - | 3 | 432 | 5323566 | ||
M79305.1 | - | 1 | 42 | |||
Z22821.1 | - | 1 | 122 |