Search
Search

Surf6| Gene Symbol: Surf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009298.3 | NP_033324.1 | 2 | 208 | ||
| GenBank mRNA | AK077989.1 | - | 2 | 210 | ||
| AK132194.1 | - | 2 | 244 | |||
| BC037634.1 | - | 2 | 255 | 4952465 | ||
| X92842.1 | - | 2 | 191 | |||