Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009387.2 | NP_033413.2 | 2, 3 | 334 | ||
GenBank mRNA | AK085188.1 | - | 2, 3 | 183 | ||
AK087988.1 | - | 2, 3 | 137 | |||
AK139244.1 | - | 2, 3 | 154 | |||
BC012215.1 | - | 2, 3 | 138 | 3969287 | ||
BC091758.1 | - | 2, 3 | 167 | 4978985 | ||
BY773001.1 | - | 2, 3 | 334 | |||
M11945.1 | - | 2, 3 | 131 | |||
M19438.1 | - | 1, 2 | 24 | |||
S57244.1 | - | 2, 3 | 131 |