Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053943.1 | - | 1 | 328 | ||
AK128980.1 | - | 1 | 404 | |||
AK158276.1 | - | 1 | 351 | |||
AK159330.1 | - | 1 | 332 | |||
AK159342.1 | - | 1 | 351 | |||
AK159377.1 | - | 1 | 322 | |||
AK159409.1 | - | 1 | 352 | |||
AK159774.1 | - | 1 | 323 | |||
AK159957.1 | - | 1 | 323 | |||
BC082577.1 | - | 1 | 340 | 30648530 |
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK177445.1 | - | 94 |