Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 1651 | ||
NM_001290351.1 | NP_001277280.1 | 4 | 1642 | |||
NM_001290353.1 | NP_001277282.1 | 4 | 1549 | |||
NM_009554.4 | NP_033580.3 | 4 | 1639 | |||
GenBank mRNA | AK032469.1 | - | 4 | 1521 | ||
BC024863.1 | - | 2 | 1262 | |||
BC063757.1 | - | 4 | 1523 | 6331110 | ||
X52533.1 | - | 1 | 937 | |||
X64413.1 | - | 4 | 1445 | |||
X89264.1 | - | 5 | 1551 |