Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 18 | 2367 | ||
NM_001308031.1 | NP_001294960.1 | 11 | 1317 | |||
NM_005246.3 | NP_005237.2 | 20 | 2753 | |||
XM_011543266.1 | XP_011541568.1 | 21 | 2651 | |||
XM_011543267.1 | XP_011541569.1 | 21 | 4535 | |||
XM_011543269.1 | XP_011541571.1 | 20 | 4389 | |||
XM_011543270.2 | XP_011541572.1 | 20 | 2566 | |||
XM_011543271.2 | XP_011541573.1 | 20 | 2606 | |||
XM_011543272.1 | XP_011541574.1 | 20 | 2447 | |||
XM_011543276.2 | XP_011541578.1 | 12 | 1569 | |||
XM_017009229.1 | XP_016864718.1 | 22 | 2798 | |||
XM_017009230.1 | XP_016864719.1 | 19 | 2521 | |||
XM_017009231.1 | XP_016864720.1 | 19 | 4344 | |||
XM_017009232.1 | XP_016864721.1 | 20 | 2439 | |||
XM_017009233.1 | XP_016864722.1 | 19 | 2382 | |||
GenBank mRNA | AK299855.1 | - | 18 | 2332 | ||
AK315234.1 | - | 18 | 2441 | |||
J03358.1 | - | 20 | 2753 | |||
JQ412173.1 | - | 11 | 1317 |