Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CORO1A SIRNA Assays ›
          Gene Symbol:
          Coro1a
          Gene Name:
          coronin, actin binding protein 1A
          Gene Aliases:
          Clabp, Lmb3, TACO, p57
          Chromosome Location:
          Chr.7: 126699774 - 126704816 on Build GRCm38
          UniGene ID:
          Mm.290482
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 66233

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Coro1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301374.1 NP_001288303.1 5, 6 883
          NM_009898.3 NP_034028.1 5, 6 796
          GenBank mRNA AF047388.1 - 4, 5 630
          AF143955.1 - 5, 6 767
          AF495468.1 - 5, 6 734
          AK040885.1 - 5, 6 740
          AK146290.1 - 5, 6 734
          AK150962.1 - 5, 6 738
          AK151755.1 - 5, 6 738
          AK155527.1 - 5, 6 739
          AK155860.1 - 5, 6 734
          AK165296.1 - 5, 6 738
          AK172447.1 - 5, 6 734
          BC002136.1 - 5, 6 724 3484185
          BC053398.1 - 5, 6 714 30076939
          U89399.1 - 1, 2 166

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagolysosome assembly
          calcium ion transport
          phagocytosis
          actin filament organization
          regulation of actin polymerization or depolymerization
          regulation of cell shape
          cell migration
          actin cytoskeleton organization
          positive regulation of cell migration
          leukocyte chemotaxis
          regulation of actin filament polymerization
          negative regulation of vesicle fusion
          cell-substrate adhesion
          uropod organization
          regulation of actin cytoskeleton organization
          response to cytokine
          nerve growth factor signaling pathway
          positive regulation of T cell proliferation
          T cell homeostasis
          natural killer cell degranulation
          negative regulation of neuron apoptotic process
          homeostasis of number of cells within a tissue
          positive regulation of T cell activation
          positive chemotaxis
          negative regulation of actin nucleation
          regulation of release of sequestered calcium ion into cytosol
          early endosome to recycling endosome transport
          cellular response to interleukin-4
          actin binding
          actin monomer binding
          protein binding
          protein C-terminus binding
          cytoskeletal protein binding
          myosin heavy chain binding
          identical protein binding
          protein homodimerization activity
          phosphatidylinositol 3-kinase binding
          poly(A) RNA binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qlhjl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0