Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRK4 SIRNA Assays ›
Gene Symbol:
GRK4
Gene Name:
G protein-coupled receptor kinase 4
Gene Aliases:
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location:
Chr.4: 2963504 - 3040760 on Build GRCh38
UniGene ID:
Hs.32959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
663
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004056.1
NP_001004056.1
2
603
NM_001004057.1
NP_001004057.1
3
699
NM_005307.2
NP_005298.2
2
603
NM_182982.2
NP_892027.2
3
699
XM_011513447.2
XP_011511749.1
3
811
XM_011513448.2
XP_011511750.1
3
811
XM_011513449.2
XP_011511751.1
2
715
XM_011513450.2
XP_011511752.1
3
811
XM_011513451.2
XP_011511753.1
3
811
XM_011513452.2
XP_011511754.1
3
811
XM_011513453.2
XP_011511755.1
3
811
XM_011513454.2
XP_011511756.1
3
811
XM_011513455.2
XP_011511757.1
3
811
XM_011513456.2
XP_011511758.1
3
811
XM_011513457.2
XP_011511759.1
3
811
XM_017008052.1
XP_016863541.1
3
811
XM_017008053.1
XP_016863542.1
3
811
XM_017008054.1
XP_016863543.1
3
811
XM_017008055.1
XP_016863544.1
2
715
XM_017008056.1
XP_016863545.1
3
811
XM_017008061.1
XP_016863550.1
2
466
XM_017008063.1
XP_016863552.1
2
466
XM_017008064.1
XP_016863553.1
2
250
XM_017008065.1
XP_016863554.1
3
811
GenBank mRNA
AK223581.1
-
2
611
AK292390.1
-
3
744
BC117320.1
-
3
587
40125871
U33054.1
-
3
498
U33055.1
-
2
402
U33056.1
-
3
498
X97879.1
-
2
246
X97880.1
-
2
246
X97881.1
-
3
342
X98118.1
-
3
342
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor internalization
protein phosphorylation
signal transduction
regulation of G-protein coupled receptor protein signaling pathway
regulation of rhodopsin mediated signaling pathway
receptor internalization
G-protein coupled receptor kinase activity
ATP binding
rhodopsin kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-zs8m7:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0