Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cx3cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009987.4 | NP_034117.3 | 2 | 2673 | ||
GenBank mRNA | AK035902.1 | - | 2 | 2649 | ||
AK045634.1 | - | 2 | 2661 | |||
BC012653.1 | - | 2 | 2630 | 4017224 |