Search
Search

Ext2| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010163.3 | NP_034293.2 | 13 | 2166 | ||
| GenBank mRNA | AK134438.1 | - | 9 | 1107 | ||
| AK148090.1 | - | 13 | 2156 | |||
| AK148369.1 | - | 12 | 1927 | |||
| AK150155.1 | - | 13 | 2158 | |||
| AK164342.1 | - | 13 | 2152 | |||
| AK179607.1 | - | 3 | 249 | |||
| AK184805.1 | - | 3 | 278 | |||
| AK188211.1 | - | 3 | 250 | |||
| BC006597.1 | - | 13 | 2139 | 3965079 | ||
| U67837.1 | - | 13 | 2141 | |||
| U72141.1 | - | 13 | 2145 | |||