Search
Search

Gli1| Gene Symbol: Gli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010296.2 | NP_034426.2 | 10 | 1097 | ||
| GenBank mRNA | AB025922.1 | - | 10 | 1107 | ||
| AF026305.1 | - | 10 | 1097 | |||
| BC131650.1 | - | 9 | 1075 | 40110429 | ||
| CV782971.1 | - | 5 | 491 | |||