Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IKBKG SIRNA Assays ›
          Gene Symbol:
          Ikbkg
          Gene Name:
          inhibitor of kappaB kinase gamma
          Gene Aliases:
          1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO
          Chromosome Location:
          Chr.X: 74393291 - 74456792 on Build GRCm38
          UniGene ID:
          Mm.12967
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 67420

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ikbkg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136067.2 NP_001129539.1 3 340
          NM_001161421.1 NP_001154893.1 3 340
          NM_001161422.1 NP_001154894.1 2 158
          NM_001161423.1 NP_001154895.1 2 210
          NM_001161424.1 NP_001154896.1 2 210
          NM_010547.2 NP_034677.2 2 236
          NM_178590.4 NP_848705.1 2 236
          GenBank mRNA AF069542.1 - 1 95
          AF513109.1 - 3 251
          AK037020.1 - 2 215
          AK148163.1 - 2 194
          AK154095.1 - 2 222
          AY112937.1 - 3 340
          BC021431.1 - 3 330 5030381
          BY768708.1 - 2 246
          CJ099503.1 - 3 325
          CN701499.1 - 2 158
          CN717929.1 - 2 151
          CV556906.1 - 2 177

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_199103, XM_008773607, XM_008773602, XM_006229585, XM_008773601, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          activation of NF-kappaB-inducing kinase activity
          positive regulation of gene expression
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of NF-kappaB transcription factor activity
          establishment of vesicle localization
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          protein binding
          protein domain specific binding
          ubiquitin protein ligase binding
          protein homodimerization activity
          peroxisome proliferator activated receptor binding
          metal ion binding
          protein heterodimerization activity
          K63-linked polyubiquitin binding
          linear polyubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0