Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIPK2 SIRNA Assays ›
          Gene Symbol:
          Hipk2
          Gene Name:
          homeodomain interacting protein kinase 2
          Gene Aliases:
          1110014O20Rik, B230339E18Rik, Stank
          Chromosome Location:
          Chr.6: 38687512 - 38876190 on Build GRCm38
          UniGene ID:
          Mm.23790
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 67532

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hipk2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136065.2 NP_001129537.1 5 1694
          NM_001294143.1 NP_001281072.1 5 1770
          NM_001294144.1 NP_001281073.1 5 1770
          NM_010433.2 NP_034563.2 5 1770
          GenBank mRNA AF077659.1 - 5 1533
          AF170301.1 - 5 1520
          AF170302.1 - 5 1520
          AF208292.1 - 5 1607
          AF273680.1 - 5 1491
          AF333791.1 - 5 1397
          AF333792.1 - 5 1397
          BC031904.1 - 5 1503 5368577

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          transforming growth factor beta receptor signaling pathway
          smoothened signaling pathway
          adult walking behavior
          positive regulation of cell proliferation
          adult locomotory behavior
          anterior/posterior pattern specification
          retina layer formation
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          neuron differentiation
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          positive regulation of protein binding
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          embryonic camera-type eye morphogenesis
          voluntary musculoskeletal movement
          positive regulation of sequence-specific DNA binding transcription factor activity
          embryonic retina morphogenesis in camera-type eye
          lens induction in camera-type eye
          SMAD protein signal transduction
          iris morphogenesis
          negative regulation of nucleic acid-templated transcription
          nucleotide binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          DNA binding
          transcription corepressor activity
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          SMAD binding
          virion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0