Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159850.1 | NP_001153322.1 | 4 | 778 | ||
NM_001281847.1 | NP_001268776.1 | 4 | 669 | |||
NM_001281848.1 | NP_001268777.1 | 3 | 510 | |||
NM_010607.3 | NP_034737.2 | 4 | 874 | |||
GenBank mRNA | AK138427.1 | - | 4 | 550 | ||
AK140716.1 | - | 4 | 549 | |||
AK164197.1 | - | 3 | 510 | |||
AY736359.1 | - | 4 | 513 | |||
BC062094.1 | - | 4 | 492 | 6848897 | ||
BY267214.1 | - | 3 | 313 | |||
U73488.2 | - | 5 | 951 |