Search
Search

Kcnj12| Gene Symbol: Kcnj12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267593.1 | NP_001254522.1 | 2 | 1944 | ||
| NM_010603.6 | NP_034733.4 | 3 | 2116 | |||
| GenBank mRNA | AK017299.1 | - | 3 | 2117 | ||
| AK032271.1 | - | 2 | 1946 | |||
| AK162389.1 | - | 1 | 1016 | |||
| X80417.1 | - | 1 | 1681 | |||