Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Klra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101620.1 | NP_001095090.1 | 4 | 546 | ||
NM_010650.3 | NP_034780.1 | 474 | ||||
GenBank mRNA | AK143236.1 | - | 475 | |||
L78253.1 | - | 392 | ||||
U12889.1 | - | 4 | 597 |
Gene Symbol: Klra14-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY860969.1 | - | 401 | |||
AY860974.1 | - | 3 | 371 |