Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANXA1 SIRNA Assays ›
          Gene Symbol:
          Anxa1
          Gene Name:
          annexin A1
          Gene Aliases:
          Anx-1, Anx-A1, C430014K04Rik, Lpc-1, Lpc1
          Chromosome Location:
          Chr.19: 20373434 - 20390671 on Build GRCm38
          UniGene ID:
          Mm.248360
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 67916

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Anxa1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010730.2 NP_034860.2 3, 4 257
          GenBank mRNA AK002681.1 - 3, 4 259
          AK140838.1 - 3, 4 260
          AK150564.1 - 3, 4 259
          AK150718.1 - 3, 4 259
          AK153156.1 - 3, 4 259
          AK153490.1 - 3, 4 259
          AK167612.1 - 3, 4 259
          BC002289.1 - 2, 3 212 3590168
          BC004594.1 - 2, 3 198 3587973
          CT009771.1 - 2, 3 183
          CT010298.1 - 2, 3 183
          EU684130.1 - 2, 3 183
          M24554.1 - 2, 3 168
          X07486.1 - 2, 3 196

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000700, BC035993, BT019896, AB451274, X05908, and others.

          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA strand renaturation
          neutrophil homeostasis
          signal transduction
          cell surface receptor signaling pathway
          peptide cross-linking
          insulin secretion
          keratinocyte differentiation
          positive regulation of vesicle fusion
          positive regulation of prostaglandin biosynthetic process
          DNA duplex unwinding
          positive regulation of neutrophil apoptotic process
          DNA rewinding
          regulation of cell proliferation
          positive regulation of apoptotic process
          alpha-beta T cell differentiation
          arachidonic acid secretion
          negative regulation of protein secretion
          cellular response to hydrogen peroxide
          prolactin secretion
          cellular response to glucocorticoid stimulus
          neutrophil clearance
          positive regulation of G1/S transition of mitotic cell cycle
          negative regulation of phospholipase A2 activity
          negative regulation of interleukin-8 secretion
          single-stranded DNA binding
          single-stranded RNA binding
          helicase activity
          phospholipase inhibitor activity
          structural molecule activity
          calcium ion binding
          protein binding
          phospholipid binding
          calcium-dependent phospholipid binding
          phospholipase A2 inhibitor activity
          protein binding, bridging
          double-stranded DNA-dependent ATPase activity
          annealing helicase activity
          protein homodimerization activity
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0