Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010773.2 | NP_034903.2 | 1 | 625 | ||
GenBank mRNA | AF072243.1 | - | 1 | 583 | ||
AF072245.1 | - | 1 | 583 | |||
AK082825.1 | - | 1 | 62 | |||
AK147008.1 | - | 1 | 510 | |||
BC026698.1 | - | 1 | 585 | |||
BC046607.1 | - | 1 | 625 | 6399206 | ||
BC057289.1 | - | 1 | 457 | |||
BC058864.1 | - | 1 | 445 |