Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDST2 SIRNA Assays ›
          Gene Symbol:
          Ndst2
          Gene Name:
          N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2
          Gene Aliases:
          Mndns, NDST-2
          Chromosome Location:
          Chr.14: 20723730 - 20734621 on Build GRCm38
          UniGene ID:
          Mm.4084
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68078

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ndst2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010811.2 NP_034941.2 3 1058
          GenBank mRNA AF074925.1 - 1 308
          AK150100.1 - 3 839
          AK170298.1 - 3 840
          AK213521.1 - 1 198
          BC050829.1 - 3 823
          BC110479.1 - 1 456 40048114
          BC110480.1 - 1 456 40048115
          U02304.1 - 2 820
          X75885.1 - 1 465

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001105740, XM_008770478, XM_008770579, XM_008770578, XM_006251623, and others.

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of angiotensin levels in blood
          glycosaminoglycan biosynthetic process
          metabolic process
          cellular process
          heparan sulfate proteoglycan biosynthetic process
          N-acetylglucosamine 6-O-sulfotransferase activity
          N-acetylgalactosamine 4-O-sulfotransferase activity
          catalytic activity
          GPI-anchor transamidase activity
          heparan sulfate 2-O-sulfotransferase activity
          sulfotransferase activity
          [heparan sulfate]-glucosamine N-sulfotransferase activity
          HNK-1 sulfotransferase activity
          transferase activity
          hydrolase activity
          heparan sulfate 6-O-sulfotransferase activity
          trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
          1-phenanthrol sulfotransferase activity
          3-phenanthrol sulfotransferase activity
          4-phenanthrol sulfotransferase activity
          trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
          9-phenanthrol sulfotransferase activity
          2-phenanthrol sulfotransferase activity
          phenanthrol sulfotransferase activity
          deacetylase activity
          1-hydroxypyrene sulfotransferase activity
          galactose 3-O-sulfotransferase activity
          proteoglycan sulfotransferase activity
          cholesterol sulfotransferase activity
          coenzyme F390-A hydrolase activity
          coenzyme F390-G hydrolase activity
          hydroxyjasmonate sulfotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0