Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321859.1 | NP_001308788.1 | 4, 5 | 2774 | ||
NM_001321860.1 | NP_001308789.1 | 4, 5 | 2844 | |||
NM_001321861.1 | NP_001308790.1 | 4, 5 | 2807 | |||
NM_001321862.1 | NP_001308791.1 | 4, 5 | 2794 | |||
NM_001321863.1 | NP_001308792.1 | 4, 5 | 2744 | |||
NM_001321904.1 | NP_001308833.1 | 5, 6 | 3222 | |||
NM_007008.2 | NP_008939.1 | 2, 3 | 464 | |||
NM_020532.4 | NP_065393.1 | 4, 5 | 3527 | |||
NM_153828.2 | NP_722550.1 | 2, 3 | 1070 | |||
NM_207520.1 | NP_997403.1 | 3, 4 | 1127 | |||
NM_207521.1 | NP_997404.1 | 4, 5 | 2785 | |||
NR_135829.1 | 2, 3 | 350 | ||||
NR_135830.1 | 2, 3 | 328 | ||||
XM_005264434.3 | XP_005264491.1 | 5, 6 | 2921 | |||
XM_017004518.1 | XP_016860007.1 | 5, 6 | 2960 | |||
XM_017004519.1 | XP_016860008.1 | 5, 6 | 3077 | |||
GenBank mRNA | AB015639.1 | - | 2, 3 | 838 | ||
AB020693.1 | - | 4, 5 | 3363 | |||
AB040462.1 | - | 4, 5 | 3473 | |||
AB040463.1 | - | 2, 3 | 1016 | |||
AB073351.1 | - | 2, 3 | 1199 | |||
AF063601.1 | - | 2, 3 | 1787 | |||
AF077050.1 | - | 2, 3 | 463 | |||
AF087901.1 | - | 2, 3 | 355 | |||
AF125103.1 | - | 2, 3 | 464 | |||
AF132047.1 | - | 2, 3 | 951 | |||
AF132048.1 | - | 2, 3 | 370 | |||
AF148537.1 | - | 4, 5 | 3370 | |||
AF148538.1 | - | 2, 3 | 913 | |||
AF177332.1 | - | 2, 3 | 369 | |||
AF320999.1 | - | 3, 4 | 2533 | |||
AF333336.1 | - | 4, 5 | 2789 | |||
AJ251383.1 | - | 4, 5 | 3229 | |||
AJ251384.1 | - | 2, 3 | 772 | |||
AJ251385.1 | - | 2, 3 | 250 | |||
AK075039.1 | - | 2, 3 | 1004 | |||
AK129806.1 | - | 2, 3 | 929 | |||
AK130812.1 | - | 2, 3 | 831 | |||
AY102276.1 | - | 2, 3 | 484 | |||
AY102277.1 | - | 2, 3 | 1016 | |||
AY102278.1 | - | 3, 4 | 1073 | |||
AY102279.1 | - | 4, 5 | 3473 | |||
AY123245.1 | - | 4, 5 | 2786 | |||
AY123246.1 | - | 4, 5 | 2844 | |||
AY123247.1 | - | 4, 5 | 2807 | |||
AY123248.1 | - | 4, 5 | 2793 | |||
AY123249.1 | - | 4, 5 | 2754 | |||
AY123250.1 | - | 4, 5 | 2744 | |||
BC001035.2 | - | 2, 3 | 435 | |||
BC007109.1 | - | 2, 3 | 361 | |||
BC010737.1 | - | 2, 3 | 777 | |||
BC012619.1 | - | 2, 3 | 927 | |||
BC014366.1 | - | 2, 3 | 361 | |||
BC016165.1 | - | 3, 4 | 1076 | |||
BC026788.1 | - | 2, 3 | 958 | |||
BC068991.1 | - | 2, 3 | 1020 | |||
BC071848.1 | - | 3, 4 | 758 | |||
BC150182.1 | - | 4, 5 | 3477 | 40148210 | ||
BC152425.1 | - | 4, 5 | 3363 | 8862539 | ||
BC152555.1 | - | 4, 5 | 3477 | 40148984 | ||
CN429712.1 | - | 2, 3 | 299 | |||
DQ778739.1 | - | 2, 3 | 250 |