Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDGFRA SIRNA Assays ›
          Gene Symbol:
          Pdgfra
          Gene Name:
          platelet derived growth factor receptor, alpha polypeptide
          Gene Aliases:
          AI115593, CD140a, Pdgfr-2
          Chromosome Location:
          Chr.5: 75151401 - 75198211 on Build GRCm38
          UniGene ID:
          Mm.221403
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68504

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pdgfra
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083316.1 NP_001076785.1 3 544
          NM_011058.2 NP_035188.2 3 527
          GenBank mRNA AK035501.1 - 3 484
          AK081664.1 - 3 526
          AK136490.1 - 3 513
          AK139094.1 - 2 759
          AK145097.1 - 3 1127
          AK147267.1 - 3 476
          AK163952.1 - 3 591
          BC020405.1 - 3 458
          BC037467.1 - 3 466
          BC053036.1 - 3 544 5704645
          M57683.1 - 3 426
          M84607.1 - 3 537

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_006206, BC015186, BC063414, AK308353, AK316578, and others.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          luteinization
          in utero embryonic development
          hematopoietic progenitor cell differentiation
          protein phosphorylation
          chemotaxis
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cytosolic calcium ion concentration
          multicellular organism development
          estrogen metabolic process
          positive regulation of cell proliferation
          female gonad development
          anatomical structure morphogenesis
          organ morphogenesis
          negative regulation of platelet activation
          positive regulation of phospholipase C activity
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          signal transduction involved in regulation of gene expression
          extracellular matrix organization
          lung development
          adrenal gland development
          positive regulation of cell migration
          male genitalia development
          Leydig cell differentiation
          cellular response to reactive oxygen species
          platelet-derived growth factor receptor-alpha signaling pathway
          cellular response to platelet-derived growth factor stimulus
          positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
          wound healing
          odontogenesis of dentin-containing tooth
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of DNA replication
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          embryonic digestive tract morphogenesis
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          regulation of chemotaxis
          cardiac myofibril assembly
          palate development
          face morphogenesis
          cell chemotaxis
          retina vasculature development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          cellular response to amino acid stimulus
          metanephric glomerular capillary formation
          regulation of mesenchymal stem cell differentiation
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          platelet-derived growth factor alpha-receptor activity
          vascular endothelial growth factor-activated receptor activity
          platelet-derived growth factor receptor binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein complex binding
          vascular endothelial growth factor binding
          protein homodimerization activity
          phosphatidylinositol 3-kinase binding
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nzvfq:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0