Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSEN2 SIRNA Assays ›
          Gene Symbol:
          Psen2
          Gene Name:
          presenilin 2
          Gene Aliases:
          ALG-3, Ad4h, Alg3, PS-2, PS2, Psnl2, STM2
          Chromosome Location:
          Chr.1: 180227004 - 180263476 on Build GRCm38
          UniGene ID:
          Mm.330850
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68971

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Psen2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128605.1 NP_001122077.1 11 1486
          NM_011183.3 NP_035313.2 11 1495
          GenBank mRNA AF038935.1 - 10 1420
          AK077576.1 - 2 940
          AK148111.1 - 11 1487
          AK154119.1 - 11 1431
          BC010403.1 - 10 1441 4038761
          BM935552.1 - 5 609
          U49111.1 - 3 387
          U57324.1 - 11 1444
          U57325.1 - 4 636

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031087, XM_006250410, XM_006250408, XM_008769877, XM_006250407, and others.

          Panther Classification:

          Molecular Function -

          aspartic protease

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell fate specification
          somitogenesis
          positive regulation of receptor recycling
          negative regulation of protein phosphorylation
          hair follicle development
          hematopoietic progenitor cell differentiation
          T cell activation involved in immune response
          myeloid leukocyte differentiation
          proteolysis
          membrane protein ectodomain proteolysis
          calcium ion transport
          negative regulation of epidermal growth factor-activated receptor activity
          regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          Notch receptor processing
          learning or memory
          memory
          embryo development
          protein transport
          protein processing
          dorsal/ventral neural tube patterning
          embryonic limb morphogenesis
          forebrain development
          negative regulation of protein complex assembly
          negative regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          endoplasmic reticulum calcium ion homeostasis
          intracellular signal transduction
          locomotion
          anagen
          amyloid precursor protein catabolic process
          positive regulation of apoptotic process
          positive regulation of catalytic activity
          regulation of protein binding
          skin morphogenesis
          cellular protein metabolic process
          regulation of synaptic plasticity
          lung alveolus development
          thymus development
          brain morphogenesis
          beta-amyloid metabolic process
          positive regulation of coagulation
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein transferase activity
          protein maturation
          cardiac muscle contraction
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic signaling pathway
          endopeptidase activity
          aspartic-type endopeptidase activity
          protein binding
          peptidase activity
          hydrolase activity
          aspartic endopeptidase activity, intramembrane cleaving

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rvxms:80/100.66.78.92:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0