Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL2 SIRNA Assays ›
          Gene Symbol:
          Ccl2
          Gene Name:
          chemokine (C-C motif) ligand 2
          Gene Aliases:
          AI323594, HC11, JE, MCAF, MCP-1, MCP1, SMC-CF, Scya2, Sigje
          Chromosome Location:
          Chr.11: 82035577 - 82037452 on Build GRCm38
          UniGene ID:
          Mm.290320
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 69169

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ccl2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011333.3 NP_035463.1 3 333
          GenBank mRNA AF065929.1 - 3 271
          AF065930.1 - 3 271
          AF065931.1 - 3 271
          AF065932.1 - 3 271
          AF065933.1 - 3 271
          AK132590.1 - 3 332
          AK150937.1 - 3 334
          AK151789.1 - 3 332
          AK153443.1 - 3 332
          AK153468.1 - 3 334
          AK153520.1 - 3 334
          BC055070.1 - 2 636
          BC145867.1 - 3 300 40131170
          BC145869.1 - 3 300 40131172
          CT010187.1 - 3 248

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of leukocyte mediated cytotoxicity
          positive regulation of endothelial cell proliferation
          leukocyte migration involved in inflammatory response
          positive regulation of leukocyte migration
          positive regulation of cellular extravasation
          cellular calcium ion homeostasis
          chemotaxis
          inflammatory response
          immune response
          cytoskeleton organization
          transforming growth factor beta receptor signaling pathway
          glial cell migration
          regulation of cell shape
          response to heat
          response to wounding
          regulation of vascular endothelial growth factor production
          positive regulation of macrophage chemotaxis
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          positive regulation of cell-cell adhesion
          neutrophil chemotaxis
          positive regulation of tumor necrosis factor production
          positive regulation of collagen biosynthetic process
          helper T cell extravasation
          monocyte homeostasis
          positive regulation of cell adhesion
          vascular endothelial growth factor receptor signaling pathway
          eosinophil chemotaxis
          macrophage chemotaxis
          lymphocyte chemotaxis
          positive regulation of synaptic transmission
          positive regulation of T cell activation
          positive regulation of nitric-oxide synthase biosynthetic process
          chemokine-mediated signaling pathway
          cellular response to lipopolysaccharide
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          positive regulation of monocyte chemotaxis
          positive regulation of immune complex clearance by monocytes and macrophages
          positive regulation of protein targeting to membrane
          positive regulation of apoptotic cell clearance
          G-protein coupled receptor binding
          cytokine activity
          protein binding
          chemokine activity
          heparin binding
          CCR2 chemokine receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0