Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011392.2 | NP_035522.2 | 13 | 2062 | ||
GenBank mRNA | AB163921.1 | - | 2067 | |||
AB163922.1 | - | 13 | 2006 | |||
AK018740.2 | - | 13 | 2017 | |||
AK032618.1 | - | 6 | 1153 | |||
BC020529.1 | - | 13 | 2005 | 4223484 | ||
L33878.1 | - | 12 | 1989 | |||
U22465.1 | - | 13 | 2006 |