Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011414.3 | NP_035544.1 | 3 | 525 | ||
GenBank mRNA | AK010170.1 | - | 3 | 312 | ||
BC028509.1 | - | 3 | 525 | 1513866 | ||
U73004.1 | - | 736 | ||||
U88093.1 | - | 3 | 302 | |||
U94341.1 | - | 3 | 303 |