Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSC2 SIRNA Assays ›
          Gene Symbol:
          Tsc2
          Gene Name:
          tuberous sclerosis 2
          Gene Aliases:
          Nafld, Tcs2
          Chromosome Location:
          Chr.17: 24595816 - 24632627 on Build GRCm38
          UniGene ID:
          Mm.30435
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 69665

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tsc2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039363.2 NP_001034452.1 2 213
          NM_001286713.1 NP_001273642.1 2 213
          NM_001286714.1 NP_001273643.1 2 208
          NM_001286716.1 NP_001273645.1 2 208
          NM_001286718.1 NP_001273647.1 2 213
          NM_001286720.1 NP_001273649.1 2 213
          NM_011647.3 NP_035777.2 2 213
          GenBank mRNA AK143799.1 - 2 214
          AK147016.1 - 2 189
          AK147302.1 - 2 153
          AK147487.1 - 2 182
          AK147874.1 - 2 178
          AK147908.1 - 2 214
          AK148062.1 - 2 205
          AK170589.1 - 2 204
          AK171007.1 - 2 185
          AK196271.1 - 1 68
          BC052449.1 - 2 189 6409037
          BC060701.1 - 2 189 6409037
          BY742714.1 - 2 217
          U37775.1 - 1 128
          U39818.1 - 2 184

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neural tube closure
          negative regulation of protein kinase activity
          protein import into nucleus
          signal transduction
          heart development
          protein localization
          negative regulation of cell proliferation
          phosphatidylinositol 3-kinase signaling
          negative regulation of phosphatidylinositol 3-kinase signaling
          cell projection organization
          regulation of endocytosis
          negative regulation of Wnt signaling pathway
          negative regulation of TOR signaling
          cellular response to insulin stimulus
          negative regulation of MAP kinase activity
          protein kinase B signaling
          positive regulation of GTPase activity
          protein transport into plasma membrane raft
          establishment of protein localization
          negative regulation of cell size
          positive regulation of transcription from RNA polymerase II promoter
          glucose import
          regulation of insulin receptor signaling pathway
          negative regulation of insulin receptor signaling pathway
          insulin-like growth factor receptor signaling pathway
          negative regulation of epithelial cell proliferation
          positive chemotaxis
          regulation of small GTPase mediated signal transduction
          protein homooligomerization
          protein heterooligomerization
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          GTPase activator activity
          protein binding
          phosphatase binding
          protein homodimerization activity
          protein heterodimerization activity
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0