Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164078.1 | NP_001157550.1 | 3, 4 | 514 | ||
NM_001164079.1 | NP_001157551.1 | 3, 4 | 508 | |||
NM_011585.4 | NP_035715.1 | 3, 4 | 508 | |||
GenBank mRNA | AK009502.1 | - | 2, 3 | 181 | ||
AK009914.1 | - | 2, 3 | 181 | |||
AK028340.1 | - | 2, 3 | 181 | |||
AK088486.1 | - | 3, 4 | 444 | |||
AK144222.1 | - | 3, 4 | 461 | |||
AK185472.1 | - | 1, 2 | 26 | |||
AK188600.1 | - | 1, 2 | 26 | |||
AK219644.1 | - | 1, 2 | 26 | |||
BC023813.1 | - | 3, 4 | 423 | 5353938 | ||
BC027826.1 | - | 3, 4 | 350 | |||
BC046812.1 | - | 3, 4 | 397 | 5702183 | ||
U00689.1 | - | 3, 4 | 449 | |||
U55862.1 | - | 3, 4 | 316 |