Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011749.4 | NP_035879.1 | 4 | 624 | ||
GenBank mRNA | AF316548.1 | - | 4 | 563 | ||
AK146591.1 | - | 4 | 486 | |||
BC026144.1 | - | 4 | 489 | 3991738 | ||
CK634070.1 | - | 2 | 272 | |||
U80078.1 | - | 1 | 148 | |||
X98096.1 | - | 4 | 521 |