Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033962.1 | NP_001029134.1 | 4 | 1057 | ||
NM_011668.2 | NP_035798.2 | 6 | 1244 | |||
NM_173010.3 | NP_766598.1 | 4 | 1059 | |||
GenBank mRNA | AK029133.1 | - | 4 | 1060 | ||
AK083795.1 | - | 4 | 988 | |||
AK220549.1 | - | 4 | 957 | |||
BC131658.1 | - | 4 | 1045 | 40110636 | ||
BC131659.1 | - | 4 | 1045 | 40110637 | ||
U82122.1 | - | 2 | 494 | |||
U96636.1 | - | 6 | 791 |