Search
Search

Rcn2| Gene Symbol: Rcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011992.2 | NP_036122.2 | 3, 4 | 667 | ||
| GenBank mRNA | AF049125.1 | - | 3, 4 | 656 | ||
| AK041996.1 | - | 664 | ||||
| AK076153.1 | - | 3, 4 | 494 | |||
| AK077486.1 | - | 3, 4 | 643 | |||
| AK078106.1 | - | 3, 4 | 635 | |||
| AK169765.1 | - | 3, 4 | 641 | |||
| BC132320.1 | - | 3, 4 | 632 | 40130597 | ||
| BC145668.1 | - | 3, 4 | 632 | 40130971 | ||