Search
Search

GRIN2B| Gene Symbol: GRIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000834.3 | NP_000825.2 | 2, 3 | 598 | ||
| XM_011520628.2 | XP_011518930.1 | 3, 4 | 1391 | |||
| XM_011520629.2 | XP_011518931.1 | 4, 5 | 1198 | |||
| XM_017019219.1 | XP_016874708.1 | 3, 4 | 946 | |||
| GenBank mRNA | BC113618.1 | - | 2, 3 | 497 | 8322670 | |
| BC113620.1 | - | 2, 3 | 497 | 8322672 | ||
| U11287.1 | - | 2, 3 | 629 | |||
| U28861.1 | - | 1, 2 | 419 | |||
| U88963.1 | - | 2, 3 | 598 | |||
| U90278.1 | - | 2, 3 | 628 | |||