Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RYR2 SIRNA Assays ›
          Gene Symbol:
          RYR2
          Gene Name:
          ryanodine receptor 2
          Gene Aliases:
          ARVC2, ARVD2, RYR-2, RyR, VTSIP
          Chromosome Location:
          Chr.1: 237042208 - 237833988 on Build GRCh38
          UniGene ID:
          Hs.109514
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7063

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RYR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001035.2 NP_001026.2 3, 4 399
          XM_006711802.3 XP_006711865.1 3, 4 593
          XM_006711803.3 XP_006711866.1 3, 4 593
          XM_006711804.3 XP_006711867.1 3, 4 593
          XM_006711805.3 XP_006711868.1 3, 4 593
          XM_006711806.3 XP_006711869.1 3, 4 593
          XM_006711807.3 XP_006711870.1 3, 4 593
          XM_006711808.3 XP_006711871.1 3, 4 593
          XM_006711809.3 XP_006711872.1 3, 4 593
          XM_006711810.3 XP_006711873.1 3, 4 593
          GenBank mRNA AB204532.1 - 1, 2 100
          X98330.1 - 3, 4 400

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of heart rate
          embryonic heart tube morphogenesis
          left ventricular cardiac muscle tissue morphogenesis
          cardiac muscle hypertrophy
          detection of calcium ion
          calcium ion transport
          cellular calcium ion homeostasis
          positive regulation of heart rate
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          response to muscle activity
          calcium-mediated signaling
          BMP signaling pathway
          response to caffeine
          ion transmembrane transport
          calcium-mediated signaling using intracellular calcium source
          response to muscle stretch
          release of sequestered calcium ion into cytosol
          positive regulation of sequestering of calcium ion
          regulation of cytosolic calcium ion concentration
          response to redox state
          regulation of cardiac muscle contraction
          cardiac muscle contraction
          canonical Wnt signaling pathway
          calcium ion transport into cytosol
          sarcoplasmic reticulum calcium ion transport
          cellular response to caffeine
          cellular response to epinephrine stimulus
          establishment of protein localization to endoplasmic reticulum
          ventricular cardiac muscle cell action potential
          Purkinje myocyte to ventricular cardiac muscle cell signaling
          cell communication by electrical coupling involved in cardiac conduction
          type B pancreatic cell apoptotic process
          positive regulation of the force of heart contraction
          regulation of AV node cell action potential
          regulation of SA node cell action potential
          regulation of atrial cardiac muscle cell action potential
          regulation of ventricular cardiac muscle cell action potential
          positive regulation of calcium-transporting ATPase activity
          regulation of cardiac conduction
          ryanodine-sensitive calcium-release channel activity
          calcium channel activity
          calcium ion binding
          protein binding
          calmodulin binding
          calcium-release channel activity
          enzyme binding
          protein kinase A catalytic subunit binding
          protein kinase A regulatory subunit binding
          identical protein binding
          protein self-association
          suramin binding
          ion channel binding
          calcium-induced calcium release activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline